missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 10515983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10515983 1 g Glass bottle 1g
10386412 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515983 Supplier Thermo Scientific Acros Supplier No. 374760010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 25026-64-6
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00973904
InChI Key QFMRANWPGGSNHS-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro-benzoic acid, 3-fluoro-5-chlorobenzoic acid, 5-chloro-3-fluorobenzoic acid, benzoic acid, 3-chloro-5-fluoro, 3-chloro-5-fluorobenzoicacid, pubchem1373, acmc-209ggm, intermediates-zcf02673, ksc497i9n, rarechem al be 1247
PubChem CID 2734840
IUPAC Name 3-chloro-5-fluorobenzoic acid
SMILES C1=C(C=C(C=C1F)Cl)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 153°C to 154°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(Cl)COOH
Quantity 1 g
Formula Weight 174.56
Percent Purity 97%
Physical Form Crystalline Powder or Granules
Chemical Name or Material 3-Chloro-5-fluorobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.