missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenol, 98%

Product Code. 11396599
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396599 1 g 1g
11306609 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396599 Supplier Thermo Scientific Alfa Aesar Supplier No. H26195.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 202982-70-5 | C6H4ClFO | 146.545 g/mol

3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorophenol is used as an intermediate in organic synthesis. It plays an important role in the preparation of substituted amides as antagonists and/or inverse agonists of cannabinoid-1 receptor, substituted N-(arylmethyl)aryloxy arylcarboxamide and heteroarylcarboxamide antagonists for the PGE2 receptor EP4 for the treatment of pain, inflammation, osteoarthritis, and rheumatoid arthritis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202982-70-5
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD00042212
InChI Key ZMFBYTAAMHWQHD-UHFFFAOYSA-N
Synonym 3-chloro-5-fluoro-phenol, phenol, 3-chloro-5-fluoro, pubchem2858, acmc-209f8b, 3-chloranyl-5-fluoranyl-phenol, phenol, 3-chloro-5-fluoro-9ci
PubChem CID 2773713
IUPAC Name 3-chloro-5-fluorophenol
SMILES C1=C(C=C(C=C1F)Cl)O

Specifications

Refractive Index 1.537
Quantity 1 g
UN Number UN3265
Solubility Information Immiscible with water.
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-fluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.