missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific™

Product Code. 15471576
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471576 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471576 Supplier Thermo Scientific Alfa Aesar Supplier No. H54775.AE
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Reacts with water.

Notes
Store at 4°C.
TRUSTED_SUSTAINABILITY

Specifications

CAS 480438-50-4
Molecular Formula C6H3BrClFMg
MDL Number MFCD02260214
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 3-chloro-5-fluorophenylmagnesium bromide, bromo 3-chloro-5-fluorophenyl magnesium, ikmssontwseqmt-uhfffaoysa-m, magnesium,bromo 3-chloro-5-fluorophenyl, magnesium, bromo 3-chloro-5-fluorophenyl, 3-chloro-5-fluorophenylmagnesium bromide solution, 3-chloro-5-fluorophenylmagnesium bromide, 0.50 m in 2-methf, 3-chloro-5-fluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3-chloro-5-fluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Reacts with water.
InChI Key IKMSSONTWSEQMT-UHFFFAOYSA-M
SMILES FC1=CC(Cl)=CC([Mg]Br)=C1
IUPAC Name magnesium;1-chloro-3-fluorobenzene-5-ide;bromide
Molecular Weight (g/mol) 233.75
PubChem CID 2778633
Formula Weight 233.75
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 3-Chloro-5-fluorophenylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.