missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-methoxybenzoic acid, 98%

Product Code. 15444607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444607 1 g 1g
15454607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444607 Supplier Thermo Scientific Alfa Aesar Supplier No. H31960.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 82477-67-6 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82477-67-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD01317994
InChI Key RRVHCVOVXPMKGF-UHFFFAOYSA-N
Synonym 3-chloro-5-methoxy-benzoic acid, 5-chloro-m-anisic acid, 3-carboxy-5-chloroanisole, 3-chloro-5-methoxybenzoicacid, benzoic acid, 3-chloro-5-methoxy, pubchem14017, acmc-20ac57, ksc447q4j, cs-waa0107, benzoicacid, 3-chloro-5-methoxy
PubChem CID 12909341
IUPAC Name 3-chloro-5-methoxybenzoic acid
SMILES COC1=CC(Cl)=CC(=C1)C(O)=O

Specifications

Quantity 1 g
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 3-Chloro-5-methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.