missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenesulfonamide, 98%

Product Code. 11348105
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348105 1 g 1g
11358105 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348105 Supplier Thermo Scientific Alfa Aesar Supplier No. L08349.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17260-71-8 | C6H6ClNO2S | 191.63 g/mol

It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can react with 1-chloro-4-isocyanato-benzene to prepare C13H10Cl2N2O3S in the presence of 1 N aq. NaOH and solvent acetone.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17260-71-8
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.63
MDL Number MFCD00051976
InChI Key WSYQJNPRQUFCGL-UHFFFAOYSA-N
Synonym benzenesulfonamide, 3-chloro, 3-chlorobenzene-1-sulfonamide, 3-chlorobenzenesulphonamide, 3-chloro-benzenesulfonamide, m-chlorobenzenesulfonamide, acmc-20a3ig, 3-chlorobenzensulfonamide, 3-chloranylbenzenesulfonamide, benzenesulfonamide,3-chloro, benzenesulfonamide, 3-chloro-9ci
PubChem CID 519377
IUPAC Name 3-chlorobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 144°C to 148°C
Quantity 1 g
Beilstein 2832245
Solubility Information Slightly soluble in water.
Formula Weight 191.64
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.