missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylideneacetone, 98%

Product Code. 11484103
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484103 25 g 25g
11474103 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484103 Supplier Thermo Scientific Alfa Aesar Supplier No. A10332.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20766-36-3 | C10H9ClO | 180.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20766-36-3
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00052851
InChI Key VWEPXSRBBXPYSM-AATRIKPKSA-N
Synonym 3e-4-3-chlorophenyl but-3-en-2-one, e-4-3-chloro-phenyl-but-3-en-2-one, 4-3-chlorophenyl but-3-en-2-one, e-4-3-chlorophenyl-but-3-en-2-one, 3-chlorobenzylideneacetone, 3-chloro-benzalacetone, 3-chlorostyrylmethyl ketone, 3-chlorobenzylidene acetone, chembl73680, e-4-3-chlorophenyl but-3-en-2-one
PubChem CID 5373975
IUPAC Name (E)-4-(3-chlorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=CC=C1)Cl

Specifications

Boiling Point 123°C to 125°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 180.64
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzylideneacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.