missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorocinnamic acid, predominantly trans, 98+%

Product Code. 11455443
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455443 10 g 10g
11465443 50 g 50g
11475443 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455443 Supplier Thermo Scientific Alfa Aesar Supplier No. A13416.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1866-38-2 | C9H7ClO2 | 182.603 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-38-2
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00004384
InChI Key FFKGOJWPSXRALK-SNAWJCMRSA-N
Synonym 3-chlorocinnamic acid, 3-3-chlorophenyl acrylic acid, m-chlorocinnamic acid, e-3-3-chlorophenyl acrylic acid, 2e-3-3-chlorophenyl acrylic acid, trans-3-chlorocinnamic acid, e-3-chlorocinnamic acid, 2e-3-3-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-3-chlorophenyl, rarechem bk hc t320
PubChem CID 735250
IUPAC Name (E)-3-(3-chlorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)Cl)C=CC(=O)O

Specifications

Melting Point 161°C to 164°C
Quantity 10 g
Beilstein 2207378
Formula Weight 182.61
Percent Purity ≥98%
Chemical Name or Material 3-Chlorocinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.