missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10476282
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10476282 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10476282 Supplier Thermo Scientific Maybridge Supplier No. SEW02033EA
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 73217-31-9
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9ClN2O2
MDL Number MFCD00084967
Quantity 10 g
Synonym 3-chloromethyl-5-4-methoxyphenyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-4-methoxyphenyl, 3-chloromethyl-5-4-methoxy-phenyl-1,2,4 oxadiazole, 1-3-chloromethyl 1,2,4-oxadiazol-5-yl-4-methoxybenzene, 3-chloromethyl-5-4-methyloxy phenyl-1,2,4-oxadiazole
InChI Key LSYCZBSPYWGGHC-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C2=NC(=NO2)CCl
IUPAC Name 3-(chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole
Molecular Weight (g/mol) 224.644
PubChem CID 2797131
Formula Weight 224.65
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-(4-methoxyphenyl)-1, 2, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.