missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)benzamide, 97%, Thermo Scientific™

Product Code. p-2698226
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10714583

Brand: Thermo Scientific Maybridge S08130CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135654-16-9
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00175948
InChI Key OOYATKQQGVPYQR-UHFFFAOYSA-N
Synonym 3-chloromethyl benzamide, benzamide,3-chloromethyl, benzamide, 3-chloromethyl-9ci, 3-chloromethyl-benzamide, 3-carbamoylbenzyl chloride, acmc-20a41t, 3-aminocarbonyl benzyl chloride, 3-carbamoylbenzyl chloride, 3-aminocarbonyl benzyl chloride
PubChem CID 737145
IUPAC Name 3-(chloromethyl)benzamide
SMILES C1=CC(=CC(=C1)CCl)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH2ClC6H4C(O)NH2
Quantity 250 mg
Formula Weight 169.61
Percent Purity 97%
Chemical Name or Material 3-(chloromethyl)benzamide
compliance-icons
Product Identifier
  • 3-(Chloromethyl)benzamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.