missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water

Product Code. 10252652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10252652 25 g Plastic Bottle 25g
10430711 100 g Plastic Bottle 100g
10023240 500 g Plastic Bottle 500g
10626661 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10252652 Supplier Thermo Scientific Acros Supplier No. 255790250
  • ONLINE EXCLUSIVE PRICE
    45.55 €
    42.50 € / 25g
    Save 3.05 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5 | CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloroperoxybenzoic Acid
Name Note 70 - 75%
CAS 937-14-4, 535-80-8, 7732-18-5
CAS Min % 70.0, 25.0, 25.0
CAS Max % 75.0, 30.0, 30.0
Melting Point 92.0°C to 94.0°C
Color White
Density 0.5600g/mL
Assay Percent Range di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC)
Packaging Plastic Bottle
Molecular Formula C7H5ClO3
Linear Formula ClC6H4CO3H
MDL Number MFCD00002127
Quantity 25 g
UN Number 3106
Beilstein 09,IV,972
Fieser 01,135; 02,68; 03,49; 04,85; 05,120; 06,110; 07,62; 08,97; 09,108; 10,92; 11,122; 12,118; 13,76; 15,86; 16,80; 17,76
Merck Index 15,2154
Specific Gravity 0.56
Synonym 3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in koh, soluble in chloroform and acetone
InChI Key NHQDETIJWKXCTC-UHFFFAOYSA-N
SMILES OOC(=O)C1=CC=CC(Cl)=C1
IUPAC Name 3-chlorobenzenecarboperoxoic acid
Molecular Weight (g/mol) 172.56
PubChem CID 70297
ChEBI CHEBI:52091
Formula Weight 172.57
Percent Purity 70-75%
Physical Form Moist Powder
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chloroperoxybenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Organic peroxide Type C & D
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.