missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, 98%

Product Code. 11449050
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449050 50 g 50g
11469050 100 g 100g
11459050 250 g 250g
11439050 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11449050 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur A13671.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol

3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.

Solubility
Soluble in ethanol and ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Spécification

Melting Point 30°C to 34°C
Density 1.245
Boiling Point 214°C
Flash Point 120°C (248°F)
Refractive Index 1.566
Quantity 50 g
UN Number UN2020
Beilstein 1634401
Merck Index 14,2154
Solubility Information Soluble in ethanol and ether.
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Chlorophenol
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.