missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, 98%

Product Code. 11469050
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469050 100 g 100g
11449050 50 g 50g
11459050 250 g 250g
11439050 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11469050 Supplier Thermo Scientific Alfa Aesar Supplier No. A13671.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol

3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is an important intermediate in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenol is widely used in the pharmaceutical, dyes and organic synthesis industry. It is also used as a disinfectant. It is an important intermediate in the synthesis of chemicals.

Solubility
Soluble in ethanol and ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Specifications

Melting Point 30°C to 34°C
Density 1.245
Boiling Point 214°C
Flash Point 120°C (248°F)
Refractive Index 1.566
Quantity 100 g
UN Number UN2020
Beilstein 1634401
Merck Index 14,2154
Solubility Information Soluble in ethanol and ether.
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Chlorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.