missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, 99%

Product Code. 10353562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10353562 5 g Glass bottle 5g
10675332 25 g Glass bottle 25g
10580181 100 g Glass bottle 100g
13418308 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10353562 Supplier Thermo Scientific Acros Supplier No. 109810050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-43-0 | C6H5ClO | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002256
InChI Key HORNXRXVQWOLPJ-UHFFFAOYSA-N
Synonym m-chlorophenol, phenol, 3-chloro, phenol, m-chloro, 3-hydroxychlorobenzene, m-chlorophenic acid, meta-chlorophenol, unii-z2z7m2ftad, 3-chloro-1-hydroxybenzene, 3-chloro-phenol, ccris 641
PubChem CID 7933
ChEBI CHEBI:38855
IUPAC Name 3-chlorophenol
SMILES OC1=CC=CC(Cl)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33.0°C to 35.0°C
Color Brown to Yellow
Density 1.2180g/mL
Boiling Point 214.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H4OH
Refractive Index 1.5650 to 1.5670
Quantity 5 g
Beilstein 06,185
Merck Index 15,2157
Specific Gravity 1.218
Solubility Information Solubility in water: 27.7g/l (20°C). Other solubilities: soluble in alcohol,ether,benzene and aq. alkali
Viscosity 11.55 Pas (25°C)
Formula Weight 128.56
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 3-Chlorophenol, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.