missing translation for 'onlineSavingsMsg'
Learn More

(3-chlorophenyl)methanol, Thermo Scientific™

Product Code. 10641635
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10641635 10 g 10g
10081834 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10641635 Supplier Thermo Scientific Maybridge Supplier No. SB01130EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 873-63-2
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
InChI Key ZSRDNPVYGSFUMD-UHFFFAOYSA-N
Synonym 3-chlorobenzyl alcohol, 3-chlorophenyl methanol, m-chlorobenzyl alcohol, benzenemethanol, 3-chloro, 3-chlorobenzylalcohol, 3-chlorophenyl methan-1-ol, m-chlorobenzylalcohol, 3-chlorobenyl alcohol, pubchem3626, acmc-209qjt
PubChem CID 70117
IUPAC Name (3-chlorophenyl)methanol
SMILES C1=CC(=CC(=C1)Cl)CO

Specifications

Density 1.2100g/mL
Assay Percent Range 97%
Linear Formula ClC6H4CH2OH
Quantity 10 g
Formula Weight 142.58
Percent Purity 97%
Chemical Name or Material (3-Chlorophenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.