missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylhydrazine hydrochloride, 97%

Product Code. 11479120
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479120 250 g 250g
11459120 10 g 10g
11469120 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479120 Supplier Thermo Scientific Alfa Aesar Supplier No. A14770.30
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2312-23-4 | C6H8Cl2N2 | 179.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2312-23-4
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.044
MDL Number MFCD00012935
InChI Key CRRIAWUJYMLJOE-UHFFFAOYSA-N
Synonym 3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
PubChem CID 75331
IUPAC Name (3-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)NN.Cl

Specifications

Melting Point ∼242°C (decomposition)
Odor Odorless
Quantity 250 g
Beilstein 3565828
Formula Weight 179.05
Percent Purity 97%
Chemical Name or Material 3-Chlorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.