missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophthalic anhydride, 95+%

Product Code. 10358663 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10358663

Brand: Thermo Scientific Acros 381290250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117-21-5 | C8H3ClO3 | 182.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-21-5
Molecular Formula C8H3ClO3
Molecular Weight (g/mol) 182.56
MDL Number MFCD00023107
InChI Key UERPUZBSSSAZJE-UHFFFAOYSA-N
Synonym 3-chlorophthalic anhydride, 4-chloroisobenzofuran-1,3-dione, 3-chlorophthalic acid anhydride, unii-vo7nxt0137, 1,3-isobenzofurandione, chloro, 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione, 4-chloranyl-2-benzofuran-1,3-dione, 3clpa, pubchem19464, 3-chlorophthalicanhydride
PubChem CID 67014
IUPAC Name 4-chloro-2-benzofuran-1,3-dione
SMILES ClC1=CC=CC2=C1C(=O)OC2=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 123°C to 126°C
Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 25 g
Formula Weight 182.56
Percent Purity 95+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Chlorophthalic anhydride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.