missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl phenyl sulfide, 97%

Product Code. 11419277
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419277 25 g 25g
11409277 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419277 Supplier Thermo Scientific Alfa Aesar Supplier No. B21345.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4911-65-3 | C9H11ClS | 186.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4911-65-3
Molecular Formula C9H11ClS
Molecular Weight (g/mol) 186.70
MDL Number MFCD00040122
InChI Key SBRLITLVSMPPFI-UHFFFAOYSA-N
Synonym 3-chloropropyl phenyl sulfide, 3-chloropropyl sulfanyl benzene, 3-chloropropyl thio benzene, 3-chloropropylphenylsulfide, acmc-20an5p, 3-phenylthiopropylchloride, 3-phenylthiopropyl chloride, 1-chloro-3-phenylthiopropane, 3-chloro-1-phenylthiopropane
PubChem CID 269606
IUPAC Name 3-chloropropylsulfanylbenzene
SMILES ClCCCSC1=CC=CC=C1

Specifications

Odor Stench
Quantity 25 g
Formula Weight 186.7
Percent Purity 97%
Chemical Name or Material 3-Chloropropyl phenyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.