missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophenol, 97%

Product Code. 11371717
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371717 25 g 25g
11351717 1 g 1g
11361717 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371717 Supplier Thermo Scientific Alfa Aesar Supplier No. L04806.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2037-31-2 | C6H5ClS | 144.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-31-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
MDL Number MFCD00004839
InChI Key CQJDYPZUDYXHLM-UHFFFAOYSA-N
Synonym 3-chlorothiophenol, m-chlorothiophenol, benzenethiol, 3-chloro, 3-chlorobenzene-1-thiol, m-chlorobenzenethiol, benzenethiol, m-chloro, 3-chloro thiophenol, 3-chloro-benzenethiol, 3-chloromercaptobenzene, m-chlorophenyl mercaptan
PubChem CID 16257
IUPAC Name 3-chlorobenzenethiol
SMILES SC1=CC=CC(Cl)=C1

Specifications

Density 1.244
Boiling Point 104°C to 105°C (35 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.6021
Quantity 25 g
UN Number UN1760
Beilstein 1854087
Sensitivity Air Sensitive
Formula Weight 144.62
Percent Purity 97%
Chemical Name or Material 3-Chlorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.