missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyclopentyloxy)-4-methoxybenzaldehyde, Thermo Scientific™

Product Code. 10769025
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10769025 25 g Amber Glass Bottle 25g
10263372 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10769025 Supplier Thermo Scientific Maybridge Supplier No. SB01923EB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67387-76-2
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
InChI Key FZFWPURYSWKIRT-UHFFFAOYSA-N
Synonym 3-cyclopentyloxy-4-methoxybenzaldehyde, 3-cyclopentyl-oxy-4-methoxybenzaldehyde, 2-cyclopentyloxy-4-formylanisole, 3-cyclopentoxy-4-methoxybenzaldehyde, 3-cyclopentyloxy-4-methoxybenzaldeyde, 3-cylopentyloxy-4-methoxybenzaldehyde, 3-cyclopentyloxy-4-methoxybenzaidehyde, 5-formyl-2-methoxyphenoxy cyclopentane
PubChem CID 2735893
IUPAC Name 3-cyclopentyloxy-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)OC2CCCC2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 3-(Cyclopentyloxy)-4-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.