missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethoxy)nitrobenzene, TRC

Product Code. 30180293
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180293 100 mg 100mg
30179906 50 mg 50mg
30180162 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180293 Supplier Toronto Research Chemicals Supplier No. TRCD452240100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22236-07-3
Molecular Formula C7H5NO3F2
Molecular Weight (g/mol) 189.11
Synonym 1-(Difluoromethoxy)-3-nitrobenzene;
IUPAC Name 1-(difluoromethoxy)-3-nitrobenzene
SMILES [O-][N+](=O)c1cccc(OC(F)F)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H5F2NO3/c8-7(9)13-6-3-1-2-5(4-6)10(11)12/h1-4,7H
Formula Weight 189.0237
Chemical Name or Material 3-(Difluoromethoxy)nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.