missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoic acid, 98%

Product Code. 11450174
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450174 500 g 500g
11430174 25 g 25g
11440174 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450174 Supplier Thermo Scientific Alfa Aesar Supplier No. A13612.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 99-64-9 | C9H11NO2 | 165.192 g/mol

3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

Solubility
Soluble in methanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-64-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00002497
InChI Key NEGFNJRAUMCZMY-UHFFFAOYSA-N
Synonym 3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
PubChem CID 66837
IUPAC Name 3-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 148°C to 153°C
Odor Odorless
Quantity 500 g
Beilstein 2208586
Solubility Information Soluble in methanol (50mg/ml). Insoluble in water.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 3-Dimethylaminobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.