missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoic acid, 99%

Product Code. 10346370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346370 25 g Plastic bottle 25g
10746141 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346370 Supplier Thermo Scientific Acros Supplier No. 115750250
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 99-64-9 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-64-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00002497
InChI Key NEGFNJRAUMCZMY-UHFFFAOYSA-N
Synonym 3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
PubChem CID 66837
IUPAC Name 3-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=CC(=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 150.0°C to 153.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CO2H
Quantity 25 g
Beilstein 14, 392
Solubility Information Solubility in water: insoluble.
Formula Weight 165.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Dimethylaminobenzoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.