missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2,4-difluorobenzoic acid, 97%

Product Code. p-7030916
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15484447

Brand: Thermo Scientific Alfa Aesar H31812.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1017779-88-2 | C9H8F2O3 | 202.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-88-2
Molecular Formula C9H8F2O3
Molecular Weight (g/mol) 202.157
MDL Number MFCD09258723
InChI Key KUOUFQUWQYELBO-UHFFFAOYSA-N
Synonym 3-ethoxy-2,4-difluorobenzoic acid, jrd
PubChem CID 46737640
IUPAC Name 3-ethoxy-2,4-difluorobenzoic acid
SMILES CCOC1=C(C=CC(=C1F)C(=O)O)F

Specifications

Melting Point 134°C to 136°C
Quantity 1 g
Formula Weight 202.15
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,4-difluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.