missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11307575
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307575 10 g 10g
11317575 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307575 Supplier Thermo Scientific Alfa Aesar Supplier No. L05721.09
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 621-51-2 | Molecular Formula: C9H10O3 | Molecular Weight: 166.176 g/mol

3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxybenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-51-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016517
InChI Key DTFQMPQJMDEWKJ-UHFFFAOYSA-N
Synonym 3-ethoxybenzoate, m-ethoxybenzoic acid, benzoic acid, 3-ethoxy, 3-ethoxy-benzoic acid, benzoic acid, m-ethoxy, 3-ethoxybenzoicacid, 3-ethyloxy benzoic acid, 3-ethoxybenzoic acid
PubChem CID 12126
ChEBI CHEBI:27990
IUPAC Name 3-ethoxybenzoic acid
SMILES CCOC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 135°C to 137°C
Quantity 10 g
Beilstein 509159
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-Ethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.