missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylamino-4-methylphenol, tech. 90%

Product Code. p-7046010
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11333366

Brand: Thermo Scientific Alfa Aesar L12768.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-37-6 | C9H13NO | 151.21 g/mol

3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-37-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002382
InChI Key CTGSQPRDMHCIMM-UHFFFAOYSA-N
Synonym 3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269
PubChem CID 8428
IUPAC Name 3-(ethylamino)-4-methylphenol
SMILES CCNC1=CC(O)=CC=C1C

Specifications

Melting Point 84°C to 92°C
Boiling Point 135°C to 140°C (3 mmHg)
Quantity 5 g
Beilstein 3241548
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Ethylamino-4-methylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.