missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-formylbenzeneboronic acid, 95%

Product Code. 15462319
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462319 1 g 1g
15472319 5 g 5g
15482319 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462319 Supplier Thermo Scientific Alfa Aesar Supplier No. H64909.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 871126-15-7 | C7H6BFO3 | 167.93 g/mol

Undergoes unprecedented tautomeric rearrangement in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undergoes unprecedented tautomeric rearrangement in solution.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871126-15-7
Molecular Formula C7H6BFO3
Molecular Weight (g/mol) 167.93
MDL Number MFCD10697421
InChI Key GVHWLCYABLWGIR-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylphenyl boronic acid, 3-fluoro-2-formylbenzeneboronic acid, boronic acid, 3-fluoro-2-formylphenyl, acmc-209qf3, 2-formyl-3-fluorophenylboranic acid, 3-fluoro-2-formyl-phenyl boronic acid, 3-fluoro-2-formylphenylboronic acid, boronic acid, b-3-fluoro-2-formylphenyl, contains varying amounts of anhydride
PubChem CID 53412038
IUPAC Name (3-fluoro-2-formylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)C=O)(O)O

Specifications

Melting Point 123°C to 127°C
Quantity 1 g
UN Number UN3261
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 167.93
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-formylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.