missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methylaniline, 98+%

Product Code. p-7026652
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11469787

Brand: Thermo Scientific Alfa Aesar B22764.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 443-86-7 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-86-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007760
InChI Key SLDLVGFPFFLYBM-UHFFFAOYSA-N
Synonym 3-fluoro-o-toluidine, 2-fluoro-6-aminotoluene, 2-amino-6-fluorotoluene, 6-amino-2-fluorotoluene, 3-fluoro-2-methyl-aniline, benzenamine, 3-fluoro-2-methyl, 3-fluoro-2-methylphenylamine, 2-methyl-3-fluoroaniline, hydrochloride, 3-fluor-2-methylanilin
PubChem CID 285553
IUPAC Name 3-fluoro-2-methylaniline
SMILES CC1=C(N)C=CC=C1F

Specifications

Melting Point 7°C
Density 1.099
Boiling Point 205°C
Flash Point 86°C (186°F)
Odor Characteristic
Refractive Index 1.542
Quantity 5 g
UN Number UN2810
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-2-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.