missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxyphenylacetic acid, 98%, Thermo Scientific™

Product Code. 10244954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244954 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244954 Supplier Thermo Scientific Acros Supplier No. 208560010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 458-09-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00004348
InChI Key YRFBZAHYMOSSGX-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyphenylacetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, benzeneacetic acid, 3-fluoro-4-hydroxy, 2-3-fluoro-4-hydroxy-phenyl acetic acid, 4-carboxymethyl-2-fluorophenol, 3-fluoro-4-hydroxy-phenyl-acetic acid, acmc-1ahjh, opera_id_1839
PubChem CID 68014
IUPAC Name 2-(3-fluoro-4-hydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 131°C to 135°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(OH)CH2CO2H
Quantity 1 g
Beilstein 10, III, 440
Merck Index 10, 4079
Formula Weight 170.14
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-hydroxyphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoro-4-hydroxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.