missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxyaniline, 98+%

Product Code. 11438527
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438527 1 g 1g
11448527 5 g 5g
11379489 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438527 Supplier Thermo Scientific Alfa Aesar Supplier No. A18606.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 366-99-4 | C7H8FNO | 141.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-99-4
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.145
MDL Number MFCD00075040
InChI Key LJWAPDSCYTZUJU-UHFFFAOYSA-N
Synonym 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine
PubChem CID 581110
IUPAC Name 3-fluoro-4-methoxyaniline
SMILES COC1=C(C=C(C=C1)N)F

Specifications

Melting Point 80°C to 84°C
Boiling Point 135°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2811
Beilstein 2717083
Formula Weight 141.15
Percent Purity ≥98%
Chemical Name or Material 3-Fluoro-4-methoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.