missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzyl alcohol, 98%

Product Code. 11300186
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
11300186 1 g 1g
11390176 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 11300186

Brand: Thermo Scientific Alfa Aesar H32350.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96047-32-4 | C8H9FO2 | 156.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96047-32-4
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00070642
InChI Key HHWYYUUOGAUCKX-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl alcohol, 3-fluoro-4-methoxyphenyl methanol, 3-fluoro-4-methoxybenzylalcohol, 2-fluoro-4-hydroxymethyl anisole, 3-fluoro-4-methoxy-phenyl methanol, benzenemethanol,3-fluoro-4-methoxy, 3-fluoranyl-4-methoxy-phenyl methanol, 3-fluoro-4-methoxyphenyl methan-1-ol
PubChem CID 2774550
IUPAC Name (3-fluoro-4-methoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)F

Specifications

Quantity 1 g
Formula Weight 156.15
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.