missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzylamine, 97%, Thermo Scientific™

Product Code. 11312469
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312469 10 g 10g
11302469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312469 Supplier Thermo Scientific Alfa Aesar Supplier No. H26607.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-4-methoxybenzylamine is used as an intermediate in organic synthesis.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with oxidizing agents, bases, acids and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123652-95-9
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.172
MDL Number MFCD04116360
InChI Key AZHAJNNLBSKSOB-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxyphenyl methanamine, 3-fluoro-4-methoxybenzylamine, 2-fluoro-4-aminomethyl anisole, 3-fluoro-4-methoxybenzyl amine, 1-3-fluoro-4-methoxyphenyl methanamine, acmc-20an3s, 3-fluoro-4-methoxybenzenemethaneamine, 3-fluoro-4-methoxyphenyl methylamine, benzenemethanamine,3-fluoro-4-methoxy
PubChem CID 14392588
IUPAC Name (3-fluoro-4-methoxyphenyl)methanamine
SMILES COC1=C(C=C(C=C1)CN)F

Specifications

Refractive Index 1.528
Quantity 10 g
UN Number UN2735
Sensitivity Air sensitive; May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 155.17
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.