missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid, 97%
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Specifications
Specifications
| Melting Point | 161°C to 163°C |
| Quantity | 250 mg |
| Formula Weight | 212.97 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction