missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid, 97%

Product Code. p-7034909
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15449945

Brand: Alfa Aesar™ H52610.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 913835-58-2
Molecular Formula C8H9BFNO4
Molecular Weight (g/mol) 212.971
MDL Number MFCD08436059
InChI Key RZYYOLUTTNFCAU-UHFFFAOYSA-N
Synonym n-methoxy 4-borono-2-fluorobenzamide, 3-fluoro-4-methoxycarbamoyl phenyl boronic acid, 3-fluoro-4-methoxycarbamoyl benzeneboronic acid, 3-fluoro-4-methoxycarbamoyl phenylboronic acid, acmc-209r9n, 3-fluoro-4-methoxycarbamoyl phenyl boronicacid, 3-fluoranyl-4-methoxycarbamoyl phenyl boronic acid, 3-fluoro-4-methoxyamino-oxomethyl phenyl boronic acid
PubChem CID 44717475
IUPAC Name [3-fluoro-4-(methoxycarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)NOC)F)(O)O

Specifications

Melting Point 161°C to 163°C
Quantity 250 mg
Formula Weight 212.97
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(methoxycarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.