missing translation for 'onlineSavingsMsg'
Learn More

3-fluoro-4-(n-morpholinomethyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 10316814
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316814 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10316814 Supplier Thermo Scientific Maybridge Supplier No. MAY00064DA
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1073354-74-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C17H25BFNO3
MDL Number MFCD09027076
Quantity 1 g
Synonym 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-n-morpholinomethyl phenylboronic acid pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzeneboronic acid, pinacol ester, 3-fluoro-4-morpholin-4-ylmethyl benzene boronic acid, pinacol ester, 4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzyl morpholine
InChI Key QINFDPZSZHPUSC-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(CN2CCOCC2)C=C1
IUPAC Name 4-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
Molecular Weight (g/mol) 321.20
PubChem CID 43811063
Formula Weight 321.2
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(N-morpholinomethyl)phenylboronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.