missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-iodoaniline, 96%

Product Code. 15447407
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447407 1 g 1g
15467407 250 mg 250mg
15457407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447407 Supplier Thermo Scientific Alfa Aesar Supplier No. H34457.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 660-49-1 | C6H5FIN | 237.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-49-1
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD04116466
InChI Key HTEPMXLKKDKDAQ-UHFFFAOYSA-N
Synonym 5-fluoro-3-iodoaniline, 3-fluoro-5-iodo-phenylamine, pubchem4458, 3-fluoro-5-iodo aniline, 3-fluoro-5-iodophenylamine, 5-fluoro-3-iodophenylamine, benzenamine,3-fluoro-5-iodo, benzenamine, 3-fluoro-5-iodo
PubChem CID 2758986
IUPAC Name 3-fluoro-5-iodoaniline
SMILES C1=C(C=C(C=C1F)I)N

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 237.02
Percent Purity 96%
Chemical Name or Material 3-Fluoro-5-iodoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.