Learn More
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
Beskrivning
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifikationer
Specifikationer
| Melting Point | 138°C to 142°C |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 197.96 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.