missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(pentafluorothio)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15486517
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486517 1 g 1g
15408575 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486517 Supplier Thermo Scientific Alfa Aesar Supplier No. H33666.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240256-92-1
Molecular Formula C7H3ClF6OS
Molecular Weight (g/mol) 284.60
MDL Number MFCD16652427
InChI Key RCKDILSECAWCTQ-UHFFFAOYSA-N
Synonym 3-fluoro-5-pentafluorosulfur benzoyl chloride, 3-fluoro-5-pentafluoro-??-sulfanyl benzoyl chloride, 3-fluoro-5-pentafluorothio benzoyl chloride
PubChem CID 66523468
IUPAC Name 3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)benzoyl chloride
SMILES FC1=CC(=CC(=C1)C(Cl)=O)S(F)(F)(F)(F)F

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 284.6
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(pentafluorothio)benzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.