missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethoxy)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15497297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497297 1 g 1g
15428905 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497297 Supplier Thermo Scientific Alfa Aesar Supplier No. H34364.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86256-47-5
Molecular Formula C8H6F4O2
Molecular Weight (g/mol) 210.128
MDL Number MFCD18783225
InChI Key WJTUZFGGSZDUKV-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethoxy benzyl alcohol, 3-fluoro-5-trifluoromethoxy phenyl methanol
PubChem CID 15582619
IUPAC Name [3-fluoro-5-(trifluoromethoxy)phenyl]methanol
SMILES C1=C(C=C(C=C1OC(F)(F)F)F)CO

Specifications

Quantity 1 g
Formula Weight 210.13
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethoxy)benzyl alcohol
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.