missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11965011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11965011 5 g 5g
11955011 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11965011 Supplier Thermo Scientific Acros Supplier No. 442600050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 161622-05-5
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00061293
InChI Key NSGKIIGVPBTOBF-UHFFFAOYSA-M
Synonym 3-fluoro-5-trifluoromethyl benzoic acid, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, 5-fluoro-3-trifluoromethyl benzoic acid, maybridge4_001933, acmc-1bwlq, intermediates-zcf02214, ksc496a7r, rarechem al bo 0634, buttpark 32\01-71
PubChem CID 519222
IUPAC Name 3-fluoro-5-(trifluoromethyl)benzoic acid
SMILES [O-]C(=O)C1=CC(F)=CC(=C1)C(F)(F)F

Specifications

Melting Point 105.0°C to 106.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoic acid
compliance-icons
Product Identifier
  • 3-Fluoro-5-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.