missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-o-xylene, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Stable under normal conditions. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Stable under normal conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.992 |
| Boiling Point | 147°C to 148°C |
| Quantity | 5 g |
| Odor | Aromatic |
| Refractive Index | 1.486 |
| UN Number | UN1993 |
| Beilstein | 2040955 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 124.16 |
| Percent Purity | 99% |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction