missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroaniline, 98+%

Product Code. 11462337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462337 500 g 500g
11442337 25 g 25g
11452337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462337 Supplier Thermo Scientific Alfa Aesar Supplier No. A14228.36
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 372-19-0 | C6H6FN | 111.119 g/mol

3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-19-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00007758
InChI Key QZVQQUVWFIZUBQ-UHFFFAOYSA-N
Synonym m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech
PubChem CID 9742
ChEBI CHEBI:27873
IUPAC Name 3-fluoroaniline
SMILES C1=CC(=CC(=C1)F)N

Specifications

Density 1.156
Boiling Point 186°C to 187°C
Flash Point 77°C (170°F)
Odor Recognizable
Refractive Index 1.544
Quantity 500 g
UN Number UN2941
Beilstein 1305471
Solubility Information Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.
Formula Weight 111.12
Percent Purity ≥98%
Chemical Name or Material 3-Fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.