missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenol, 98%

Product Code. p-3778627 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
200 mL
Packaging:
Glass bottle
Unit Size:
10mL
200mL
50mL
This item is not returnable. View return policy

Product Code. 10500931

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-20-3 | C6H5FO | 112.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-20-3
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.1
MDL Number MFCD00002254
InChI Key SJTBRFHBXDZMPS-UHFFFAOYSA-N
Synonym m-fluorophenol, phenol, 3-fluoro, 3-fluoro-phenol, phenol, m-fluoro, meta-fluorophenol, unii-t7oma38487, m-fluoro-phenol, 3-fluorphenol, 3-fluorophenol, 3-fluoro phenol
PubChem CID 9743
IUPAC Name 3-fluorophenol
SMILES C1=CC(=CC(=C1)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 10.0°C
Color Brown-Yellow to Colorless
Density 1.2400g/mL
Boiling Point 178.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4OH
Refractive Index 1.5125 to 1.5155
Quantity 10 mL
Beilstein 06, I, 97
Specific Gravity 1.24
Solubility Information Solubility in water: insoluble
Formula Weight 112.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.