missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isothiocyanate, 97+%

Product Code. 11401597
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401597 5 g 5g
11411597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401597 Supplier Thermo Scientific Alfa Aesar Supplier No. B25001.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 404-72-8 | C7H4FNS | 153.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404-72-8
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.17
MDL Number MFCD00004804
InChI Key HDBACITVPQEAGG-UHFFFAOYSA-N
Synonym 3-fluorophenyl isothiocyanate, benzene, 1-fluoro-3-isothiocyanato, m-fluorophenyl isothiocyanate, 3-fluorophenylisothiocyanate, isothiocyanic acid 3-fluorophenyl ester, isothiocyanic acid, m-fluorophenyl ester, 1-fluoro-3-isothiocyanato-benzene, 3-fluorobenzenisothiocyanate, timtec-bb sbb006700, attercop-chm at114051
PubChem CID 9834
IUPAC Name 1-fluoro-3-isothiocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=S

Specifications

Density 1.27
Boiling Point 227°C
Flash Point 85°C (185°F)
Odor Pungent
Refractive Index 1.62
Quantity 5 g
UN Number UN2922
Beilstein 2081172
Sensitivity Moisture sensitive
Formula Weight 153.18
Percent Purity ≥97%
Chemical Name or Material 3-Fluorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.