missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylacetone, 98%, Thermo Scientific™

Product Code. 10561472 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10561472 5 g Glass bottle 5g
10008731 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10561472 Supplier Thermo Scientific Acros Supplier No. 350960050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1737-19-5
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00061144
InChI Key UWCPYXSRCQVABG-UHFFFAOYSA-N
Synonym 3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
PubChem CID 137185
IUPAC Name 1-(3-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4955 to 1.4975 (20°C, 589nm)
Quantity 5 g
Formula Weight 152.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenylacetone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.