missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylboronic acid, 97%

Product Code. 10683172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10683172 5 g Glass bottle 5g
10526261 1 g Glass bottle 1g
10297503 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683172 Supplier Thermo Scientific Acros Supplier No. 345640050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 768-35-4 | C6H6BFO2 | 139.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-35-4
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00236042
InChI Key KNXQDJCZSVHEIW-UHFFFAOYSA-N
Synonym 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
PubChem CID 2733986
IUPAC Name (3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC(F)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 211.0°C to 212.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4B(OH)2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol and thf
Formula Weight 139.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.