missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylglyoxal hydrate, Thermo Scientific™

Product Code. 10250050
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10250050 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10250050 Supplier Thermo Scientific Maybridge Supplier No. MAY00104DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121247-01-6
Packaging Amber Glass Bottle
Molecular Formula C8H7FO3
Quantity 1 g
Synonym 3-fluorophenylglyoxal hydrate, 2-3-fluorophenyl-2-oxoacetaldehyde hydrate, acmc-20an3a, 3-fluorophenyl glyoxal, 3-fluoro-a-oxobenzeneacetaldehyde, benzeneacetaldehyde,3-fluoro-a-oxo, 2-3-fluorophenyl-2-oxoethanal, hydrate
InChI Key VGFHAWFFIQFSRR-UHFFFAOYSA-N
SMILES O.FC1=CC=CC(=C1)C(=O)C=O
IUPAC Name 2-(3-fluorophenyl)-2-oxoacetaldehyde;hydrate
Molecular Weight (g/mol) 170.14
PubChem CID 2783267
Formula Weight 152.12
Percent Purity 97%
Chemical Name or Material 3-fluorophenylglyoxal hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.