missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylhydrazine hydrochloride, 97%

Product Code. 10193890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193890 5 g Glass bottle 5g
10458910 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193890 Supplier Thermo Scientific Acros Supplier No. 167370050
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2924-16-5 | C6H7FN2·ClH | 162.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2924-16-5
Molecular Formula C6H7FN2·ClH
Molecular Weight (g/mol) 162.59
MDL Number MFCD00012934
InChI Key SKVGLOFWEJFQKU-UHFFFAOYSA-N
Synonym 3-fluorophenylhydrazine hydrochloride, 3-fluorophenyl hydrazine hydrochloride, 3-fluorophenylhydrazine hcl, 3-fluorophenyl hydrazine hcl, m-fluorophenylhydrazine hydrochloride, hydrazine, 3-fluorophenyl-, monohydrochloride, 1-3-fluorophenyl hydrazine hydrochloride, 3-fluoro-phenyl-hydrazine, pubchem3341, acmc-20a0dq
PubChem CID 2723914
IUPAC Name (3-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)F)NN.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 268.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H4NHNH2·HCl
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 162.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluorophenylhydrazine hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluorophenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.