missing translation for 'onlineSavingsMsg'
Learn More

3-Formyl Rifamycin, TRC

Product Code. 17904615
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904615 100 mg 100mg
17900325 1 g 1g
17920465 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904615 Supplier Toronto Research Chemicals Supplier No. TRCF700950100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-22-3
Molecular Formula C38 H47 N O13
Molecular Weight (g/mol) 725.78
Synonym Rifampicinaldehyde, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-8-carboxaldehyde, 1,2-dihydro-5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-, 21-acetate (7CI,8CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., 3-Formylrifampicin SV, 3-Formylrifamycin, 3-Formylrifamycin SV, NCI 145-635, Rifaldehyde, Rifamycin AF
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c(C=O)c(O)c4c3C2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C38H47NO13/c1-16-11-10-12-17(2)37(48)39-28-23(15-40)32(45)25-26(33(28)46)31(44)21(6)35-27(25)36(47)38(8,52-35)50-14-13-24(49-9)18(3)34(51-22(7)41)20(5)30(43)19(4)29(16)42/h10-16,18-20,24,29-30,34,42-46H,1-9H3,(H,39,48)/b11-10+,14-13+,17-12-/t16-,18+,19+,20+,24-,29-,30+,34+,38-/m0/s1
Formula Weight 725.3047
Percent Purity >95%
Chemical Name or Material 3-Formylrifamycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.