missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzaldehyde, 97%

Product Code. p-4228722 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10556040

Brand: Thermo Scientific Acros 168560010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 704-13-2 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-13-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007109
InChI Key AUBBVPIQUDFRQI-UHFFFAOYSA-N
Synonym benzaldehyde, 3-hydroxy-4-nitro, 3-hydroxy-4-nitro-benzaldehyde, 4-nitro-3-hydroxybenzaldehyde, 3-formyl-6-nitrophenol, acmc-1bm06, ksc495a6r, 3-hydroxy-4-nitro benzaldehyde, 3-hydroxy-4-nitrobenzaldehyde
PubChem CID 69712
IUPAC Name 3-hydroxy-4-nitrobenzaldehyde
SMILES OC1=CC(C=O)=CC=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 131.0°C to 134.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(NO2)CHO
Quantity 1 g
Beilstein 08,62
Formula Weight 167.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-nitrobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-4-nitrobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.