missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Citalopram, TRC

Product Code. 30011516
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011516 10 mg 10mg
30008513 1 mg 1mg
30010338 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011516 Supplier Toronto Research Chemicals Supplier No. TRCH91835010MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 411221-53-9
Molecular Formula C20 H21 F N2 O2
Molecular Weight (g/mol) 340.39
Synonym 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-hydroxy-, 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-hydroxy-5-isobenzofurancarbonitrile, 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroiso-benzofuran-5-carbonitrile, 3-Hydroxy Citalopram, 3-Hydroxycitalopram
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-3H-2-benzofuran-5-carbonitrile
SMILES CN(C)CCCC1(OC(O)c2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H21FN2O2/c1-23(2)11-3-10-20(15-5-7-16(21)8-6-15)18-9-4-14(13-22)12-17(18)19(24)25-20/h4-9,12,19,24H,3,10-11H2,1-2H3
Formula Weight 340.1587
Chemical Name or Material 3-hydroxy Citalopram
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.